4-Acetyl-2-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Acetyl-2-methylphenyl)boronic acid

A263408-100MG A263408-1G A263408-250MG
77193-896EA 35.02 USD
77193-896 77193-898 77193-900
4-Acetyl-2-methylphenylboronic acid 97%
4-Acetyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MW: 177.993 g/mol
MDL Number: MFCD19237178
CAS Number: 2096331-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR