(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene) 95%

Supplier: Ambeed
Warning

A263243-100G A263243-1G A263243-25G A263243-5G
77193-888EA 240 USD
77193-888 77193-890 77193-892 77193-894
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene) 95%
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)

Precaución: for Research Use Only

Formula: C₃₆H₅₄N₂O₂
MDL Number: MFCD00191801
CAS Number: 135616-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR