(R)-7-(3-Aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Besifloxacin HCl
A263240-10MG
A263240-1MG
77193-884EA
46.35
USD
77193-884
77193-886
(R)-7-(3-Aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 98%
(R)-7-(3-Aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₉H₂₂Cl₂FN₃O₃ MW: 430.30 g/mol |
MDL Number:
MFCD14636632 CAS Number: 405165-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...