Fmoc-(N-γ-4-methyltrityl)-D-α,γ-diaminobutyric acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((diphenyl(p-tolyl)methyl)amino)butanoic acid

A263092-100MG A263092-1G A263092-250MG
77166-232EA 8 USD
77166-232 77166-234 77166-236
Fmoc-(N-γ-4-methyltrityl)-D-α,γ-diaminobutyric acid 95%
Fmoc-(N-gamma-4-methyltrityl)-D-alpha,gamma-diaminobutyric acid

Precaución: for Research Use Only

Formula: C₃₉H₃₆N₂O₄
MDL Number: MFCD02094100
CAS Number: 1217809-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR