2-Pyridylacetic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl

A263007-10G A263007-1G A263007-25G A263007-5G A263007-100G
77193-874EA 28.84 USD
77193-874 77193-876 77193-878 77193-880 77616-104
2-Pyridylacetic acid hydrochloride 95%
2-Pyridylacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 173.60 g/mol
MDL Number: MFCD00012812
CAS Number: 16179-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR