1-(4-Fluorophenyl)propan-2-ol 96%

Supplier: Ambeed
Warning

A262943-100MG A262943-250MG
77193-870EA 23 USD
77193-870 77193-872
1-(4-Fluorophenyl)propan-2-ol 96%
1-(4-Fluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MDL Number: MFCD11934388
CAS Number: 444308-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR