1-Decyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A262869-1G A262869-25G A262869-5G
77193-864EA 5 USD
77193-864 77193-866 77193-868
1-Decyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate 98%
1-Decyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₂₇F₃N₂O₃S
MW: 372.44 g/mol
MDL Number: MFCD12022375
CAS Number: 412009-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR