2-(2-Bromophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A262746-100MG A262746-1G
77166-224EA 45 USD
77166-224 77166-226
2-(2-Bromophenyl)cyclopropanecarboxylic acid 95%
2-(2-Bromophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MDL Number: MFCD12068266
CAS Number: 767359-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR