Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A262687-100MG A262687-250MG
77166-220EA 72.1 USD
77166-220 77166-222
Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
MDL Number: MFCD25973573
CAS Number: 1335053-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR