2,6-Dichloro-4-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A262578-100MG A262578-1G A262578-250MG A262578-5G
77166-210EA 31.93 USD
77166-210 77166-212 77166-214 77166-216
2,6-Dichloro-4-hydroxybenzaldehyde 95%
2,6-Dichloro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MDL Number: MFCD06797919
CAS Number: 60964-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR