(S)-2-(1-Aminopropyl)-5-fluoro-3-phenylquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

A262279-100MG A262279-1G A262279-250MG A262279-5G
77166-202EA 13.39 USD
77166-202 77166-204 77166-206 77166-208
(S)-2-(1-Aminopropyl)-5-fluoro-3-phenylquinazolin-4(3H)-one 95%
(S)-2-(1-Aminopropyl)-5-fluoro-3-phenylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆FN₃O
MW: 297.33 g/mol
MDL Number: MFCD22124614
CAS Number: 870281-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR