Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%

Supplier: Ambeed
Warning

Synonyms: (R)-1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

A262215-100MG A262215-250MG A262215-5G A262215-1G
77193-854EA 48.41 USD
77193-854 77193-856 77571-616 77746-440
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.283 g/mol
MDL Number: MFCD12547038
CAS Number: 1799811-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR