1,9-Dibromononane 98%

Supplier: Ambeed
Warning

Synonyms: Nonamethylene dibromide

A261925-1000G A261925-100G A261925-10G A261925-25G A261925-500G A261925-5G
77193-834EA 370 USD
77193-834 77193-836 77193-838 77193-840 77193-842 77193-844
1,9-Dibromononane 98%
1,9-Dibromononane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₇CH₂Br
MDL Number: MFCD00000279
CAS Number: 4549-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR