1-(4-Iodophenyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

A261577-1G A261577-250MG
77193-830EA 90.64 USD
77193-830 77193-832
1-(4-Iodophenyl)-1H-pyrazole 98%
1-(4-Iodophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇IN₂
MW: 270.0700 g/mol
MDL Number: MFCD02681941
CAS Number: 368869-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR