Diethyl indole-2,6-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 1H-indole-2,6-dicarboxylate

A261523-100MG A261523-250MG A261523-1G
77166-194EA 17 USD
77166-194 77166-196 77612-508
Diethyl indole-2,6-dicarboxylate 97%
Diethyl indole-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD03411566
CAS Number: 107516-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR