(Tetrahydro-2H-pyran-3-yl)methanamine hydrochloride 98%

Supplier: Ambeed
Warning

A261177-100MG A261177-1G A261177-250MG A261177-5G
77166-176EA 13 USD
77166-176 77166-178 77166-180 77166-182
(Tetrahydro-2H-pyran-3-yl)methanamine hydrochloride 98%
(Tetrahydro-2H-pyran-3-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD08448168
CAS Number: 1159599-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR