AZD-5363 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, AZD 5363, AZD5363, (S)-4-amino-N-(1-(4-chlorophenyl)-3-hydroxypropyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, Capivasertib, AZD-5363
A260907-1MG
A260907-5MG
A260907-100MG
A260907-10MG
A260907-250MG
A260907-50MG
77166-172EA
28.84
USD
77166-172
77166-174
77612-496
77612-498
77612-500
77612-502
AZD-5363 98%
AZD-5363
Precaución: for Research Use Only
Formula:
C₂₁H₂₅ClN₆O₂ MW: 428.92 g/mol |
MDL Number:
MFCD22628785 CAS Number: 1143532-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...