1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95%

Supplier: Ambeed
Warning

A260871-1G A260871-250MG
77193-826EA 156.56 USD
77193-826 77193-828
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95%
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₃N₃O
MW: 247.2200 g/mol
MDL Number: MFCD01311906
CAS Number: 401930-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR