(9,9-Diphenylfluoren-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (9,9-Diphenyl-9H-fluoren-4-yl)boronic acid

A260833-250MG A260833-5G
77193-822EA 6.18 USD
77193-822 77193-824
(9,9-Diphenylfluoren-4-yl)boronic acid 97%
(9,9-Diphenylfluoren-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₂₅H₁₉BO₂
MW: 362.228 g/mol
MDL Number: MFCD29077672
CAS Number: 1224976-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR