(R)-4-(Pyrrolidin-3-yl)thiomorpholine 1,1-dioxide dihydrochloride 97%

Supplier: Ambeed
Warning

A260824-100MG A260824-1G A260824-250MG
77193-816EA 19 USD
77193-816 77193-818 77193-820
(R)-4-(Pyrrolidin-3-yl)thiomorpholine 1,1-dioxide dihydrochloride 97%
(R)-4-(Pyrrolidin-3-yl)thiomorpholine 1,1-dioxide dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂O₂S
MDL Number: MFCD28023891
CAS Number: 1015443-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR