(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)sulfonyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A260715-1G A260715-250MG A260715-5G
77166-166EA 208 USD
77166-166 77166-168 77166-170
(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)sulfonyl)phenyl)boronic acid 98+%
(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)sulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₆S
MDL Number: MFCD28384269
CAS Number: 486422-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR