Isoxazolo[4,5-c]pyridin-3-amine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A260525-100MG A260525-250MG
77166-164EA 130.81 USD
77166-164 77683-795
Isoxazolo[4,5-c]pyridin-3-amine 95%, Ambeed.Inc
Isoxazolo[4,5-c]pyridin-3-amine

Precaución: For research use only.

Formula: C₆H₅N₃O
MW: 135.12 g/mol
MDL Number: MFCD19205838
CAS Number: 1229383-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR