1-(1-Chloro-ethyl)-4-fluoro-benzene 95%

Supplier: Ambeed
Danger

A260292-100MG A260292-1G A260292-250MG A260292-5G
77193-802EA 55 USD
77193-802 77193-804 77193-806 77193-808
1-(1-Chloro-ethyl)-4-fluoro-benzene 95%
1-(1-Chloro-ethyl)-4-fluoro-benzene

Precaución: for Research Use Only

Formula: C₈H₈ClF
MDL Number: MFCD06376752
CAS Number: 456-16-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR