3-Bromo-5-formylbenzoic acid 98%

Supplier: Ambeed
Warning

A259647-1G A259647-250MG A259647-5G
77193-796EA 84.46 USD
77193-796 77193-798 77193-800
3-Bromo-5-formylbenzoic acid 98%
3-Bromo-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.027 g/mol
MDL Number: MFCD22544371
CAS Number: 398119-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR