1,3-Di-tert-butylbenzene 95%

Supplier: Ambeed
Warning

A259493-1G A259493-5G A259493-10G
77193-786EA 25 USD
77193-786 77193-788 77746-425
1,3-Di-tert-butylbenzene 95%
1,3-Di-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₂
MW: 190.33
MDL Number: MFCD00008830
CAS Number: 1014-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR