1,3-Bis(tosyloxy)propane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propane-1,3-diyl bis(4-methylbenzenesulphonate), Trimethylene Glycol Ditosylate, Trimethylene Glycol Bis(p-toluenesulphonate), 1,3-Bis(p-toluenesulphonyloxy)propane, 1,3-Propanediol Ditosylate
A259226-1G
A259226-25G
A259226-5G
A259226-100G
77193-780EA
6.18
USD
77193-780
77193-782
77193-784
77746-424
1,3-Bis(tosyloxy)propane 98%
1,3-Bis(tosyloxy)propane
Precaución: for Research Use Only
Formula:
C₁₇H₂₀O₆S₂ MW: 384.46 g/mol |
MDL Number:
MFCD00013392 CAS Number: 5469-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...