2-Amino-4,6-dichloropyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A258964-10G A258964-1G A258964-250MG A258964-5G
77166-148EA 664.35 USD
77166-148 77166-150 77166-152 77166-154
2-Amino-4,6-dichloropyrimidine-5-carbonitrile 95%
2-Amino-4,6-dichloropyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₄
MW: 189.002 g/mol
MDL Number: MFCD20727436
CAS Number: 1277179-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR