2-(1H-Indol-2-yl)aniline 97%
Supplier: Ambeed
|
Warning
|
A257993-100MG
A257993-250MG
A257993-1G
77166-144EA
30.9
USD
77166-144
77166-146
77683-786
2-(1H-Indol-2-yl)aniline 97%
2-(1H-Indol-2-yl)aniline
Precaución: For research use only.
Formula:
C₁₄H₁₂N₂ MW: 208.25 g/mol |
MDL Number:
MFCD00134432 CAS Number: 32566-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...