2,2,2,4'-Tetrafluoroacetophenone 98%

Supplier: Ambeed
Danger

A257892-10G A257892-1G A257892-25G A257892-5G
77193-768EA 30.9 USD
77193-768 77193-770 77193-772 77193-774
2,2,2,4'-Tetrafluoroacetophenone 98%
2,2,2,4'-Tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.1104 g/mol
MDL Number: MFCD00042393
CAS Number: 655-32-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR