AG-221 (Enasidenib) 98%

Supplier: Ambeed
Warning

Synonyms: Enasidenib, 2-methyl-1-((4-(6-(trifluoromethyl)pyridin-2-yl)-6-((2-(trifluoromethyl)pyridin-4-yl)amino)-1,3,5-triazin-2-yl)amino)propan-2-ol

A257789-10MG A257789-1MG A257789-5MG
77193-762EA 28.84 USD
77193-762 77193-764 77193-766
AG-221 (Enasidenib) 98%
AG-221 (Enasidenib)

Precaución: for Research Use Only

Formula: C₁₉H₁₇F₆N₇O
MW: 437.38 g/mol
MDL Number: MFCD29472245
CAS Number: 1446502-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR