1,3,4,6-Tetrathiapentalene-2,5-dione 97%

Supplier: Ambeed
Danger

Synonyms: [1,3]Dithiolo[4,5-d][1,3]dithiole-2,5-dione

A257751-1G A257751-250MG A257751-5G
77193-748EA 36.05 USD
77193-748 77193-750 77193-752
1,3,4,6-Tetrathiapentalene-2,5-dione 97%
1,3,4,6-Tetrathiapentalene-2,5-dione

Precaución: for Research Use Only

Formula: C₄O₂S₄
MW: 208.28 g/mol
MDL Number: MFCD00010403
CAS Number: 64394-45-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR