Methyl 1-oxo-2,3-dihydro-1H-indene-2-carboxylate 95%

Supplier: Ambeed
Warning

A257742-10G A257742-1G A257742-250MG
77166-134EA 697.31 USD
77166-134 77166-136 77166-138
Methyl 1-oxo-2,3-dihydro-1H-indene-2-carboxylate 95%
Methyl 1-oxo-2,3-dihydro-1H-indene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₃
MW: 190.20 g/mol
MDL Number: MFCD00667641
CAS Number: 22955-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR