Benzyl 4-(2-hydroxyethyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A257729-1G A257729-250MG
77193-744EA 344.02 USD
77193-744 77193-746
Benzyl 4-(2-hydroxyethyl)piperazine-1-carboxylate 97%
Benzyl 4-(2-hydroxyethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₃
MW: 264.32 g/mol
MDL Number: MFCD12639008
CAS Number: 14000-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR