2-(Methylsulfonylamino)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Methylsulphonamido)phenyl)boronic acid

A257723-250MG A257723-5G A257723-1G
77166-130EA 30.9 USD
77166-130 77166-132 77683-785
2-(Methylsulfonylamino)benzeneboronic acid 98%
2-(Methylsulfonylamino)benzeneboronic acid

Precaución: For research use only.

Formula: C₇H₁₀BNO₄S
MW: 215.03 g/mol
MDL Number: MFCD02179471
CAS Number: 756520-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR