(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-[bis(2-methylphenyl)phosphino]ethyl]ferrocene 99%

Supplier: Ambeed
Warning

A257714-100MG A257714-1G A257714-250MG
77166-124EA 85.49 USD
77166-124 77166-126 77166-128
(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-[bis(2-methylphenyl)phosphino]ethyl]ferrocene 99%
(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-[bis(2-methylphenyl)phosphino]ethyl]ferrocene

Precaución: for Research Use Only

Formula: C₃₄H₄₄FeP₂
MW: 570.52 g/mol
MDL Number: MFCD08561172
CAS Number: 849924-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR