2-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A257324-1G A257324-250MG A257324-5G
77166-112EA 54.59 USD
77166-112 77166-114 77166-116
2-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
2-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₅
MW: 277.08 g/mol
MDL Number: MFCD22493668
CAS Number: 1268163-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR