1-Boc-piperazine 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

A256884-1000G A256884-100G A256884-500G A256884-5G
77166-102EA 127 USD
77166-102 77166-104 77166-106 77166-108
1-Boc-piperazine 98%
1-Boc-piperazine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD00075265
CAS Number: 57260-71-6
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR