3-Bromo-4-iodophenol 98%

Supplier: Ambeed
Danger

A256627-100MG A256627-1G A256627-250MG A256627-5G A256627-25G
77166-094EA 43.26 USD
77166-094 77166-096 77166-098 77166-100 77568-486
3-Bromo-4-iodophenol 98%
3-Bromo-4-iodophenol

Precaución: For research use only.

Formula: C₆H₄BrIO
MW: 298.904 g/mol
MDL Number: MFCD17000017
CAS Number: 1037298-05-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR