4-Ethylpiperazin-2-one hydrochloride 97%

Supplier: Ambeed
Warning

A256214-1G A256214-250MG
77193-736EA 169.95 USD
77193-736 77193-738
4-Ethylpiperazin-2-one hydrochloride 97%
4-Ethylpiperazin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD22422017
CAS Number: 65464-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR