N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 95%

Supplier: Ambeed
Warning

Synonyms: 5'-O-DMT-Bz-rA

A255449-100G A255449-10G A255449-1G A255449-5G A255449-25G
77166-076EA 302.99 USD
77166-076 77166-078 77166-080 77166-082 77568-478
N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 95%
N-(9-((2R,3R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide

Precaución: For research use only.

Formula: C₃₈H₃₅N₅O₇
MW: 673.71 g/mol
MDL Number: MFCD00797466
CAS Number: 81246-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR