2-Bromo-4-iodo-5-cyanopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4-iodonicotinonitrile

A255208-100MG A255208-1G A255208-250MG
77166-070EA 87.55 USD
77166-070 77166-072 77166-074
2-Bromo-4-iodo-5-cyanopyridine 95%
2-Bromo-4-iodo-5-cyanopyridine

Precaución: for Research Use Only

Formula: C₆H₂BrIN₂
MW: 308.902 g/mol
MDL Number: MFCD16657907
CAS Number: 1061357-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR