5-Bromo-2-nitropyridin-3-ol 96%

Supplier: Ambeed
Warning

A254316-100MG A254316-250MG A254316-25G
77166-046EA 24.72 USD
77166-046 77166-048 77166-050
5-Bromo-2-nitropyridin-3-ol 96%
5-Bromo-2-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₃
MW: 218.99 g/mol
MDL Number: MFCD15527395
CAS Number: 691872-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR