CO-1686 (Rociletinib) ≥98%

Supplier: Ambeed
Warning

A254041-10MG A254041-1G A254041-1MG A254041-50MG A254041-5MG
77166-034EA 12 USD
77166-034 77166-036 77166-038 77166-040 77166-042
CO-1686 (Rociletinib) ≥98%
CO-1686 (Rociletinib)

Precaución: for Research Use Only

Formula: C₂₇H₂₈F₃N₇O₃
MDL Number: MFCD26793864
CAS Number: 1374640-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR