5-Bromo-2-chloro-1-fluoro-3-methoxybenzene 97%

Supplier: Ambeed
Warning

A253785-100MG A253785-250MG A253785-25G A253785-5G A253785-1G
77166-026EA 10.3 USD
77166-026 77166-028 77166-030 77612-442 77683-762
5-Bromo-2-chloro-1-fluoro-3-methoxybenzene 97%
5-Bromo-2-chloro-1-fluoro-3-methoxybenzene

Precaución: For research use only.

Formula: C₇H₅BrClFO
MW: 239.47 g/mol
MDL Number: MFCD28142798
CAS Number: 1261216-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR