5,6-Diamino-1,3-diethyluracil hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Diamino-1,3-diethylpyrimidine-2,4(1H,3H)-dione hydrochloride

A253433-100MG A253433-250MG
77166-014EA 8 USD
77166-014 77166-016
5,6-Diamino-1,3-diethyluracil hydrochloride 98%
5,6-Diamino-1,3-diethyluracil hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄N₄O₂·HCl
MW: 234.68 g/mol
MDL Number: MFCD22573561
CAS Number: 1785764-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR