6-Bromo-7-methyl-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

A253214-100MG A253214-250MG
77193-694EA 101.97 USD
77193-694 77193-696
6-Bromo-7-methyl-1H-pyrrolo[3,2-c]pyridine 97%
6-Bromo-7-methyl-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.06 g/mol
MDL Number: MFCD11845519
CAS Number: 1082040-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR