2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride 97%

Supplier: Ambeed

Synonyms: (2-Fluoro-6-(trifluoromethyl)phenyl)methanamine hydrochloride, 1-[2-Fluoro-6-(trifluoromethyl)phenyl]methanamine hydrochloride

A253026-100MG A253026-250MG
77166-008EA 15.45 USD
77166-008 77166-010
2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride 97%
2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₇F₄N·ClH
MW: 229.602 g/mol
MDL Number: MFCD08437622
CAS Number: 643088-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR