3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-4-(trifluoromethyl)benzoic acid

A252800-1G A252800-25G
77193-680EA 12.36 USD
77193-680 77193-682
3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene 98%
3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
MDL Number: MFCD00274359
CAS Number: 116965-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR