5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 95%

Supplier: Ambeed
Danger

A252780-100MG A252780-5G
77165-998EA 14 USD
77165-998 77892-465
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 95%
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₄
MDL Number: MFCD08448159
CAS Number: 837430-14-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR