4-(1,3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid 97%

Supplier: Ambeed
Warning

A252457-100G A252457-25G A252457-500G A252457-5G
77193-670EA 107 USD
77193-670 77193-672 77193-674 77193-676
4-(1,3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid 97%
4-(1,3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₅
MDL Number: MFCD01457304
CAS Number: 40732-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR